Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009690.2 | NP_033820.2 | 3 | 388 | ||
GenBank mRNA | AF011428.1 | - | 3 | 294 | ||
AF018268.1 | - | 3 | 327 | |||
AF018269.1 | - | 3 | 313 | |||
AK159132.1 | - | 3 | 337 | |||
AK159181.1 | - | 3 | 391 | |||
AK159823.1 | - | 3 | 356 | |||
BC006799.1 | - | 3 | 329 | 3152586 | ||
BC094459.1 | - | 3 | 341 | 4161650 |