Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aprt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009698.2 | NP_033828.2 | 3 | 286 | ||
GenBank mRNA | AB033539.1 | - | 3 | 269 | ||
AB033540.1 | - | 3 | 269 | |||
AK153201.1 | - | 3 | 251 | |||
AK182364.1 | - | 3 | 181 | |||
BC005667.1 | - | 3 | 224 | 3152733 | ||
BY115322.1 | - | 3 | 286 | |||
M11310.1 | - | 2187 |