Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009706.2 | NP_033836.2 | 4 | 4207 | ||
GenBank mRNA | CF742308.1 | - | 4 | 473 | ||
CO433888.1 | - | 3 | 500 |