Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009712.3 | NP_033842.3 | 5 | 1022 | ||
GenBank mRNA | AK083309.1 | - | 5 | 1006 | ||
AK140163.1 | - | 5 | 1072 | |||
AK154098.1 | - | 5 | 942 | |||
AK158312.1 | - | 5 | 1023 | |||
AK190270.1 | - | 2 | 84 | |||
AK205135.1 | - | 2 | 84 | |||
AK211718.1 | - | 1 | 33 | |||
BC150661.1 | - | 5 | 979 | 9087572 | ||
X92096.1 | - | 4 | 515 |