Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ate1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029895.3 | NP_001025066.1 | 5 | 766 | ||
NM_001136054.2 | NP_001129526.1 | 5 | 498 | |||
NM_001271343.1 | NP_001258272.1 | 5 | 766 | |||
NM_013799.3 | NP_038827.2 | 5 | 498 | |||
GenBank mRNA | AK138277.1 | - | 5 | 417 | ||
AK148442.1 | - | 5 | 466 | |||
AK158613.1 | - | 5 | 768 | |||
AK167591.1 | - | 5 | 458 | |||
BC042601.1 | - | 5 | 457 | 4503326 | ||
BC050948.1 | - | 5 | 448 | 6826691 | ||
DQ093639.1 | - | 5 | 358 | |||
DQ093640.1 | - | 5 | 358 |