Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ate1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029895.3 | NP_001025066.1 | 7 | 1231 | ||
NM_013799.3 | NP_038827.2 | 7 | 963 | |||
GenBank mRNA | AK077471.1 | - | 3 | 382 | ||
AK148442.1 | - | 7 | 931 | |||
AK167591.1 | - | 7 | 923 | |||
BC042601.1 | - | 7 | 922 | 4503326 | ||
DQ093639.1 | - | 7 | 823 |