Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atoh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007500.4 | NP_031526.1 | 1 | 814 | ||
GenBank mRNA | AK082354.1 | - | 1 | 837 | ||
BC010820.1 | - | 1 | 814 | 4218223 | ||
BC051256.1 | - | 1 | 832 | 6530849 |