Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007503.3 | NP_031529.2 | 3 | 149 | ||
NM_052823.2 | NP_439888.1 | 3 | 222 | |||
GenBank mRNA | AA396010.1 | - | 3 | 170 | ||
AI316264.1 | - | 3 | 133 | |||
AK002215.1 | - | 3 | 137 | |||
AK002228.1 | - | 3 | 150 | |||
AK007105.1 | - | 1 | 24 | |||
AY626243.1 | - | 3 | 271 | |||
AY626244.1 | - | 3 | 222 | |||
AY626245.1 | - | 3 | 307 | |||
BU756853.1 | - | 4 | 404 | |||
CA491597.1 | - | 3 | 388 | |||
CB600439.1 | - | 3 | 182 | |||
CB600958.1 | - | 3 | 357 |