Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007503.3 | NP_031529.2 | 4, 5 | 240 | ||
NM_052823.2 | NP_439888.1 | 4, 5 | 313 | |||
GenBank mRNA | AA396010.1 | - | 4, 5 | 261 | ||
AI316264.1 | - | 4, 5 | 224 | |||
AK002215.1 | - | 4, 5 | 228 | |||
AK002228.1 | - | 4, 5 | 241 | |||
AK007105.1 | - | 2, 3 | 115 | |||
AY626243.1 | - | 4, 5 | 362 | |||
AY626244.1 | - | 4, 5 | 313 | |||
AY626245.1 | - | 4, 5 | 398 | |||
BC024614.1 | - | 4, 5 | 229 | 4222609 | ||
BC069892.1 | - | 3, 4 | 1371 | |||
BU756853.1 | - | 2, 3 | 313 | |||
CA491597.1 | - | 4, 5 | 479 | |||
CB600439.1 | - | 4, 5 | 273 | |||
CB600958.1 | - | 4, 5 | 448 | |||
X70060.1 | - | 3, 4 | 151 |