Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007508.5 | NP_031534.2 | 11, 12 | 1353 | ||
GenBank mRNA | AK140873.1 | - | 11, 12 | 1329 | ||
AK149833.1 | - | 11, 12 | 1368 | |||
AK150742.1 | - | 11, 12 | 1339 | |||
AK152785.1 | - | 1, 2 | 67 | |||
AK153403.1 | - | 11, 12 | 1354 | |||
AK154869.1 | - | 11, 12 | 1354 | |||
AK160792.1 | - | 11, 12 | 1368 | |||
AK166857.1 | - | 11, 12 | 1343 | |||
AK170721.1 | - | 11, 12 | 1354 | |||
AK204323.1 | - | 1, 2 | 25 |