Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077265.2 | NP_001070733.1 | 5 | 958 | ||
NM_001077266.2 | NP_001070734.1 | 4 | 901 | |||
NM_001077267.2 | NP_001070735.1 | 4 | 901 | |||
NM_007516.3 | NP_031542.2 | 5 | 958 | |||
GenBank mRNA | AK077409.1 | - | 5 | 946 | ||
BC011172.1 | - | 5 | 633 | |||
BC049098.1 | - | 5 | 958 | 5717286 | ||
CA451481.1 | - | 3 | 239 | |||
CF751545.1 | - | 2 | 109 | |||
U11273.1 | - | 4 | 738 |