Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Barx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007526.4 | NP_031552.2 | 1 | 158 | ||
GenBank mRNA | AJ297677.1 | - | 1 | 185 |