Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Barx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007526.4 | NP_031552.2 | 4 | 783 | ||
GenBank mRNA | AB221538.1 | - | 3 | 381 | ||
AJ297677.1 | - | 4 | 810 | |||
BC119047.1 | - | 3 | 462 | |||
BC119049.1 | - | 3 | 462 | |||
BF011658.1 | - | 1 | 580 | |||
Y07960.1 | - | 4 | 525 |