Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007537.1 | NP_031563.1 | 3 | 244 | ||
GenBank mRNA | AF030769.1 | - | 3 | 244 | ||
AK004680.1 | - | 3 | 274 | |||
AK013244.1 | - | 3 | 261 | |||
AK015644.1 | - | 3 | 197 | |||
AK172925.1 | - | 1 | 71 | |||
BC040369.1 | - | 2 | 165 | 4511215 | ||
U59746.1 | - | 1 | 66 |