Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007537.1 | NP_031563.1 | 3 | 468 | ||
GenBank mRNA | AF030769.1 | - | 3 | 468 | ||
AK004680.1 | - | 3 | 498 | |||
AK013244.1 | - | 3 | 485 | |||
AK015644.1 | - | 3 | 421 | |||
AK172925.1 | - | 1 | 295 | |||
AY170344.3 | - | 3 | 449 | |||
BC040369.1 | - | 2 | 389 | 4511215 | ||
U59746.1 | - | 1 | 290 |