Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009777.2 | NP_033907.1 | 3 | 400 | ||
GenBank mRNA | AK140712.1 | - | 3 | 357 | ||
AK152764.1 | - | 3 | 419 | |||
AK153124.1 | - | 3 | 292 | |||
BC067001.1 | - | 3 | 336 | 5715633 | ||
M22531.1 | - | 3 | 400 | |||
M36293.1 | - | 3 | 400 | |||
X16874.1 | - | 2 | 255 |