Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007574.2 | NP_031600.2 | 3 | 575 | ||
GenBank mRNA | AK138974.1 | - | 577 | |||
BC043945.1 | - | 3 | 555 | 4972540 | ||
BC054443.1 | - | 3 | 546 | 6475447 | ||
BC075724.1 | - | 3 | 554 | 3675763 | ||
X66295.1 | - | 3 | 541 |