Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hyou1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021395.4 | NP_067370.3 | 23 | 2765 | ||
GenBank mRNA | AF228709.1 | - | 22 | 2652 | ||
AK128937.1 | - | 23 | 2712 | |||
AK145857.1 | - | 23 | 2713 | |||
AK157949.1 | - | 23 | 2683 | |||
AK171769.1 | - | 23 | 2713 | |||
BC050107.1 | - | 22 | 2670 | 30012694 | ||
BC058255.1 | - | 17 | 3476 | |||
S78797.1 | - | 14 | 1611 |