Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacybp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009786.2 | NP_033916.1 | 3, 4 | 567 | ||
GenBank mRNA | AK013924.1 | - | 3, 4 | 498 | ||
AK089043.1 | - | 3, 4 | 421 | |||
AK145013.1 | - | 3, 4 | 534 | |||
AK212050.1 | - | 1, 2 | 31 | |||
AK215280.1 | - | 1, 2 | 31 | |||
BC025948.1 | - | 3, 4 | 452 | 5150537 | ||
U97327.1 | - | 3, 4 | 397 |