Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacybp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009786.2 | NP_033916.1 | 6 | 847 | ||
GenBank mRNA | AK013924.1 | - | 6 | 778 | ||
AK089043.1 | - | 6 | 701 | |||
AK145013.1 | - | 6 | 814 | |||
AK215280.1 | - | 4 | 311 | |||
BC025948.1 | - | 6 | 732 | 5150537 | ||
U97327.1 | - | 6 | 677 |