Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009788.4 | NP_033918.1 | 11 | 864 | ||
GenBank mRNA | AK002635.1 | - | 11 | 866 | ||
AK005081.1 | - | 11 | 863 | |||
AK005243.1 | - | 11 | 864 | |||
AK028050.1 | - | 11 | 865 | |||
AK028327.1 | - | 11 | 863 | |||
AK038856.1 | - | 864 | ||||
AK048517.1 | - | 11 | 865 | |||
AK077499.1 | - | 11 | 863 | |||
AK078358.1 | - | 11 | 862 | |||
BC016421.1 | - | 11 | 795 | 4527000 | ||
M21531.1 | - | 11 | 844 | |||
M23663.1 | - | 8 | 426 |