Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042534.3 | NP_001035999.1 | 5, 6 | 769 | ||
NM_001271395.1 | NP_001258324.1 | 5, 6 | 762 | |||
NM_001271415.1 | NP_001258344.1 | 6, 7 | 759 | |||
NM_007599.4 | NP_031625.2 | 5, 6 | 616 | |||
GenBank mRNA | AK146622.1 | - | 5, 6 | 620 | ||
AK149857.1 | - | 5, 6 | 566 | |||
AK152818.1 | - | 5, 6 | 594 | |||
AK159577.1 | - | 5, 6 | 569 | |||
AK159736.1 | - | 5, 6 | 571 | |||
AK165153.1 | - | 5, 6 | 583 | |||
AK165586.1 | - | 3, 4 | 412 | |||
BC003480.1 | - | 5, 6 | 570 | 2811050 | ||
BC023101.1 | - | 5, 6 | 558 | 3983574 | ||
X54511.1 | - | 6, 7 | 758 |