Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007621.2 | NP_031647.1 | 3 | 310 | ||
GenBank mRNA | AK081816.1 | - | 3 | 281 | ||
BC010758.1 | - | 3 | 270 | 4219349 | ||
BY118518.1 | - | 3 | 311 | |||
D26123.1 | - | 3 | 297 | |||
X07411.1 | - | 3 | 267 |