Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007625.2 | NP_031651.2 | 5 | 657 | ||
GenBank mRNA | BC018304.1 | - | 5 | 688 | ||
BC045162.1 | - | 5 | 641 | |||
BC115843.1 | - | 2 | 334 | |||
BC115844.1 | - | 1 | 549 | |||
BC117801.1 | - | 5 | 531 | 40092282 | ||
BC117802.1 | - | 5 | 562 | 40092283 |