Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284508.1 | NP_001271437.1 | 4 | 489 | ||
NM_031161.4 | NP_112438.1 | 3 | 326 | |||
GenBank mRNA | AI842212.1 | - | 1 | 378 | ||
AI851721.1 | - | 1 | 378 | |||
AK002677.1 | - | 3 | 294 | |||
BC028487.1 | - | 4 | 434 | 1400830 | ||
BY127153.1 | - | 3 | 295 | |||
BY262295.1 | - | 3 | 320 | |||
BY268487.1 | - | 3 | 327 | |||
M11739.1 | - | 2 | 65 |