Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009844.2 | NP_033974.2 | 10 | 1368 | ||
GenBank mRNA | AK089835.1 | - | 10 | 1367 | ||
AK163608.1 | - | 8 | 1865 | |||
AK172008.1 | - | 10 | 1330 |