Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007641.5 | NP_031667.1 | 6, 7 | 776 | ||
GenBank mRNA | AK017903.1 | - | 6, 7 | 777 | ||
AK041428.1 | - | 6, 7 | 775 | |||
AK080040.1 | - | 6, 7 | 742 | |||
AK080148.1 | - | 774 | ||||
AK156033.1 | - | 775 | ||||
AK166571.1 | - | 6, 7 | 870 | |||
AK166659.1 | - | 6, 7 | 870 | |||
AK171631.1 | - | 6, 7 | 805 | |||
AK171638.1 | - | 6, 7 | 728 | |||
AK171659.1 | - | 6, 7 | 761 | |||
BC028322.1 | - | 6, 7 | 757 | 4503841 | ||
CT010253.1 | - | 5, 6 | 657 | |||
M62541.1 | - | 834 |