Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039150.1 | NP_001034239.1 | 14 | 2020 | ||
NM_001039151.1 | NP_001034240.1 | 9 | 1375 | |||
NM_001177785.1 | NP_001171256.1 | 16 | 2251 | |||
NM_001177786.1 | NP_001171257.1 | 12 | 1774 | |||
NM_001177787.1 | NP_001171258.1 | 11 | 1672 | |||
NM_009851.2 | NP_033981.2 | 19 | 2620 | |||
GenBank mRNA | AJ251594.1 | - | 19 | 2407 | ||
AK045226.1 | - | 9 | 1221 | |||
AK144125.1 | - | 9 | 1178 | |||
AK152099.1 | - | 9 | 1171 | |||
AK154411.1 | - | 9 | 1166 | |||
AK162720.1 | - | 9 | 1172 | |||
AK166320.1 | - | 9 | 1159 | |||
BC005676.1 | - | 9 | 1154 | |||
BC051388.1 | - | 14 | 1850 | 4910789 | ||
CT010324.1 | - | 14 | 1701 | |||
J05163.1 | - | 9 | 1037 | |||
M27129.1 | - | 9 | 1082 | |||
M27130.1 | - | 9 | 1088 | |||
M30655.1 | - | 9 | 1156 | |||
X66081.1 | - | 11 | 1356 | |||
X66082.1 | - | 12 | 1458 | |||
X66083.1 | - | 14 | 1704 | |||
X66084.1 | - | 16 | 1935 |