Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009854.2 | NP_033984.1 | 2 | 266 | ||
GenBank mRNA | AK136444.1 | - | 2 | 220 | ||
AK154339.1 | - | 2 | 221 | |||
BC024376.1 | - | 2 | 207 | 4985794 | ||
CT010368.1 | - | 2 | 153 | |||
D10329.1 | - | 2 | 206 |