Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd79a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007655.3 | NP_031681.2 | 2 | 124 | ||
GenBank mRNA | AK143605.1 | - | 2 | 125 | ||
BC027633.1 | - | 2 | 121 | 1332396 | ||
X13450.1 | - | 2 | 122 |