Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009864.2 | NP_033994.1 | 15 | 2477 | ||
GenBank mRNA | AK076369.1 | - | 15 | 2479 | ||
BC098501.1 | - | 15 | 2466 | 30023851 | ||
X06115.1 | - | 15 | 2418 | |||
X06339.1 | - | 12 | 1831 |