Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009864.2 | NP_033994.1 | 3 | 388 | ||
GenBank mRNA | AK076369.1 | - | 3 | 389 | ||
AK166267.1 | - | 3 | 389 | |||
BC098501.1 | - | 3 | 377 | 30023851 | ||
X06115.1 | - | 3 | 329 |