Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019707.4 | NP_062681.2 | 10, 11 | 1631 | ||
GenBank mRNA | AB022100.1 | - | 10, 11 | 1640 | ||
AK028002.1 | - | 2, 3 | 443 | |||
AK032409.1 | - | 10, 11 | 1662 | |||
AK039438.1 | - | 10, 11 | 1653 | |||
AK048724.1 | - | 10, 11 | 1631 | |||
AK134649.1 | - | 10, 11 | 1590 |