Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009875.4 | NP_034005.2 | 1 | 773 | ||
GenBank mRNA | AK046676.1 | - | 1 | 508 | ||
AK047669.1 | - | 1 | 774 | |||
AK050240.1 | - | 1 | 760 | |||
AK158411.1 | - | 1 | 753 | |||
AK162146.1 | - | 1 | 493 | |||
BC014296.1 | - | 1 | 756 | 4193493 | ||
CT010382.1 | - | 1 | 254 | |||
U09968.1 | - | 1 | 254 | |||
U10440.1 | - | 1 | 471 |