Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cebpz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024806.2 | NP_001019977.1 | 15 | 3014 | ||
GenBank mRNA | AK017537.1 | - | 2 | 113 | ||
AK145374.1 | - | 15 | 2996 | |||
AK158771.1 | - | 15 | 2991 | |||
AK159081.1 | - | 15 | 3011 | |||
U19891.1 | - | 15 | 2984 | |||
U19892.1 | - | 15 | 2698 |
Gene Symbol: Cebpzos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177402.1 | NP_001170873.1 | 5 | 890 | ||
NM_001177403.1 | NP_001170874.1 | 5 | 1058 |