Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cebpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007679.4 | NP_031705.3 | 1 | 729 | ||
GenBank mRNA | AK053190.1 | - | 1 | 729 | ||
AK149723.1 | - | 194 | ||||
AK151958.1 | - | 1 | 730 | |||
AK156762.1 | - | 1 | 730 | |||
BC139293.1 | - | 1 | 729 | 8862315 | ||
X61800.1 | - | 1 | 1005 |