Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007692.6 | NP_031718.1 | 5 | 962 | ||
GenBank mRNA | AB011000.1 | - | 5 | 934 | ||
AB011001.1 | - | 3 | 1284 | |||
AK164379.1 | - | 2 | 1513 | |||
AK170077.1 | - | 4 | 643 | |||
BC068195.1 | - | 5 | 480 | 4164116 | ||
BC069925.1 | - | 3 | 1283 |