Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ovgp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007696.2 | NP_031722.1 | 9, 10 | 1029 | ||
GenBank mRNA | AY521455.1 | - | 9, 10 | 1015 | ||
BC137995.1 | - | 9, 10 | 1120 | 8861014 | ||
D32137.1 | - | 9, 10 | 1029 |