Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cideb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009894.3 | NP_034024.2 | 2 | 248 | ||
GenBank mRNA | AF041377.1 | - | 2 | 236 | ||
BC012664.1 | - | 2 | 212 | 4218866 | ||
BC046340.1 | - | 2 | 242 | 6478230 | ||
CB600615.1 | - | 2 | 225 |