Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Socs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271603.1 | NP_001258532.1 | 2 | 456 | ||
NM_009896.2 | NP_034026.1 | 2 | 336 | |||
GenBank mRNA | AB000677.1 | - | 1 | 180 | ||
AB000710.1 | - | 1 | 234 | |||
AF120490.1 | - | 2 | 309 | |||
AF180302.1 | - | 2 | 314 | |||
AK028632.1 | - | 337 | ||||
AK154706.1 | - | 353 | ||||
BC132366.1 | - | 2 | 333 | 40130643 | ||
BC132368.1 | - | 2 | 333 | 40130645 | ||
U88325.1 | - | 2 | 298 |