Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013491.2 | NP_038519.1 | 10, 11 | 1259 | ||
GenBank mRNA | AK036735.1 | - | 10, 11 | 1261 | ||
AK161489.1 | - | 3, 4 | 224 | |||
AY046403.1 | - | 10, 11 | 1084 | |||
AY046404.1 | - | 11, 12 | 1163 | |||
BC114336.1 | - | 10, 11 | 1224 | 40048966 |