Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn4-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035804.1 | - | 11, 12 | 2260 | ||
AK050833.1 | - | 1, 2 | 31 | |||
AK088317.1 | - | 8, 9 | 3646 | |||
AK133759.1 | - | 12, 13 | 2194 | |||
AK209939.1 | - | 1, 2 | 42 | |||
BC005553.1 | - | 12, 13 | 2228 | 3586154 | ||
BC110668.1 | - | 12, 13 | 2620 | 6391053 | ||
DQ186662.1 | - | 10, 11 | 2118 | |||
Z49916.1 | - | 12, 13 | 2531 |