Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clns1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023671.2 | NP_076160.1 | 2 | 246 | ||
GenBank mRNA | AK011691.1 | - | 2 | 230 | ||
AK011789.1 | - | 2 | 234 | |||
AK031877.1 | - | 2 | 228 | |||
AK078050.1 | - | 2 | 202 | |||
AK133981.1 | - | 2 | 238 | |||
AK151356.1 | - | 2 | 232 | |||
AK151944.1 | - | 2 | 232 | |||
AK169259.1 | - | 2 | 210 | |||
AK180965.1 | - | 2 | 110 | |||
BC005575.1 | - | 2 | 212 | 3967933 | ||
CX236388.1 | - | 2 | 246 | |||
U53455.1 | - | 2 | 158 |