Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016674.4 | NP_057883.1 | 2 | 518 | ||
GenBank mRNA | AF072127.1 | - | 2 | 460 | ||
AK028428.1 | - | 2 | 516 | |||
AK036762.1 | - | 2 | 490 | |||
AK036780.1 | - | 2 | 519 | |||
AK040604.1 | - | 2 | 519 | |||
AK054207.1 | - | 2 | 463 | |||
AK081601.1 | - | 2 | 516 | |||
BC002003.1 | - | 2 | 471 | 3491319 | ||
BC008536.1 | - | 2 | 419 | |||
CT010299.1 | - | 2 | 305 | |||
EU076705.1 | - | 2 | 305 |