Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163432.1 | NP_001156904.1 | 4, 5 | 790 | ||
NM_007712.3 | NP_031738.2 | 4, 5 | 793 | |||
GenBank mRNA | AF033564.1 | - | 4, 5 | 515 | ||
BC015080.1 | - | 4, 5 | 778 | 3995512 | ||
DT924403.1 | - | 2, 3 | 104 |