Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111110.2 | NP_001104580.1 | 8 | 1116 | ||
NM_001284519.1 | NP_001271448.1 | 9 | 1278 | |||
NM_001284520.1 | NP_001271449.1 | 7 | 771 | |||
NM_007717.5 | NP_031743.3 | 7 | 771 | |||
GenBank mRNA | AB061276.1 | - | 8 | 1100 | ||
AB061277.1 | - | 9 | 1262 | |||
AK083394.1 | - | 5 | 614 | |||
AK145110.1 | - | 6 | 919 | |||
AK145199.1 | - | 7 | 772 | |||
AK166486.1 | - | 7 | 772 | |||
BC028474.1 | - | 8 | 1126 | |||
BC055079.1 | - | 6 | 768 | 30077348 | ||
D21826.1 | - | 6 | 743 |