Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009917.5 | NP_034047.2 | 2 | 1114 | ||
GenBank mRNA | AK141906.1 | - | 2 | 1085 | ||
AK154595.1 | - | 2 | 1115 | |||
AK155628.1 | - | 2 | 1088 | |||
AK155632.1 | - | 2 | 1086 | |||
AK155867.1 | - | 2 | 1079 | |||
AK170303.1 | - | 2 | 1085 | |||
BC103574.1 | - | 2 | 1053 | 40045779 | ||
BC103586.1 | - | 2 | 1053 | 40045781 | ||
BC103587.1 | - | 2 | 1053 | 40045780 | ||
BC105643.1 | - | 2 | 1054 | 40045777 | ||
D83648.1 | - | 2 | 1084 | |||
U47036.1 | - | 2 | 1043 |