Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnga3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282010.1 | NP_001268939.1 | 8 | 2068 | ||
NM_009918.2 | NP_034048.1 | 6 | 2013 | |||
GenBank mRNA | AJ243933.1 | - | 6 | 2022 | ||
BC035272.1 | - | 6 | 1957 | 5357087 | ||
BC049145.1 | - | 6 | 1956 | 5357087 |