Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009919.2 | NP_034049.2 | 3, 4 | 364 | ||
GenBank mRNA | AF022811.1 | - | 3, 4 | 355 | ||
AK002293.1 | - | 3, 4 | 332 | |||
AK011200.1 | - | 3, 4 | 317 | |||
AK152432.1 | - | 3, 4 | 315 | |||
AK154479.1 | - | 3, 4 | 315 | |||
AK168154.1 | - | 3, 4 | 315 | |||
BC034868.1 | - | 3, 4 | 324 | 2938052 | ||
DV653213.1 | - | 3, 4 | 418 |