Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009922.4 | NP_034052.3 | 7 | 775 | ||
GenBank mRNA | AK035548.1 | - | 7 | 748 | ||
BC138863.1 | - | 7 | 707 | 8861885 | ||
BC138864.1 | - | 7 | 707 | 8861886 | ||
BC145446.1 | - | 6 | 795 | |||
Z19542.1 | - | 7 | 769 |